Skip to Content
Merck
Sodium glycochenodeoxycholate ≥97.0% (TLC)

Sodium glycochenodeoxycholate

Sodium glycochenodeoxycholate ≥97.0% (TLC)
Synonym(s):
3α,7α-Dihydroxy-5β-cholanoic acid N-(carboxymethyl)amide sodium salt, Glycochenodeoxycholic acid sodium salt, N-(3α,7α-Dihydroxy-24-oxocholan-24-yl)glycine sodium salt
Empirical Formula (Hill Notation):
C26H42NNaO5
CAS Number:
Molecular Weight:
471.61

Products

Product Category
Brand
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing