Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium deuteroxide solution
Synonym(s):
Sodium deuteroxide solution, Sodium hydroxide-d
Linear Formula:
NaOD / D
2
O
CAS Number:
14014-06-3
Molecular Weight:
41.00
Products
Product Category
bases (3)
stable isotopes (3)
Formula Weight
Physical Form
Isotope
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
372072
Sodium deuteroxide solution,
40 wt. % in D
2
O, 99.5 atom % D
Expand
View Pricing
176788
Sodium deuteroxide solution,
40 wt. % in D
2
O, 99 atom % D
Expand
View Pricing
164488
Sodium deuteroxide solution,
30 wt. % in D
2
O, 99 atom % D
Expand
View Pricing