Skip to Content
Products
Cart
0
AR
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Sodium citrate tribasic dihydrate
Synonym(s):
Citric acid trisodium salt dihydrate, Trisodium citrate dihydrate
Linear Formula:
HOC(COONa)(CH
2
COONa)
2
· 2H
2
O
CAS Number:
6132-04-3
Molecular Weight:
294.10
EC Number:
200-675-3
Products
Product Category
buffers (4)
building blocks (1)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Special Grade
ACS reagent (2)
Puriss (1)
Puriss P.A. (1)
Purum (1)
Purum P.A. (1)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
71402
Sodium citrate tribasic dihydrate,
BioUltra, Molecular Biology
Expand
View Pricing
S4641
Sodium citrate tribasic dihydrate,
ACS reagent, ≥99.0%
Expand
View Pricing
C8532
Sodium citrate tribasic dihydrate,
Molecular Biology, ≥99%
Expand
View Pricing
C7254
Sodium citrate tribasic dihydrate,
≥98%
Expand
View Pricing
71405
Sodium citrate tribasic dihydrate,
puriss. p.a., ACS reagent, ≥99.0% (NT)
Expand
View Pricing
71406
Sodium citrate tribasic dihydrate,
purum p.a., ≥99.0% (NT)
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
Español - ES
English - EN
Learn More