Skip to Content
Merck
Silybin Pharmaceutical Secondary Standard; Certified Reference Material

Silybin

Silybin Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Silibinin, 2,3-Dihydro-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-6-(3,5,7-trihydroxy-4-oxobenzopyran-2-yl)benzodioxin, Silybin
Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade