Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silver-copper alloy, Ag85Cu15
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GF40542049
Silver-copper alloy, Ag85Cu15,
foil, 10m coil, 1mm coil width, 0.007mm thickness, as rolled
Expand
View Pricing
GF16415119
Silver-copper alloy, Ag85Cu15,
foil, 5m coil, 1mm coil width, 0.007mm thickness, as rolled
Expand
View Pricing
GF45067631
Silver-copper alloy, Ag85Cu15,
foil, 0.5m coil, 1mm coil width, 0.007mm thickness, as rolled
Expand
View Pricing
GF34957109
Silver-copper alloy, Ag85Cu15,
foil, 2m coil, 1mm coil width, 0.007mm thickness, as rolled
Expand
View Pricing
GF30067714
Silver-copper alloy, Ag85Cu15,
foil, 1m coil, 1mm coil width, 0.007mm thickness, as rolled
Expand
View Pricing