![Sildenafil for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/123/271/87accf44-a048-4fd2-8830-3dc0a415d88b/640/87accf44-a048-4fd2-8830-3dc0a415d88b.png)
Sildenafil for peak identification
![Sildenafil for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/123/271/87accf44-a048-4fd2-8830-3dc0a415d88b/640/87accf44-a048-4fd2-8830-3dc0a415d88b.png)
Synonym(s):
Sildenafil citrate salt, 1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methylpiperazine citrate salt, 5-[2-Ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-4H-pyrazolo[5,4-e]pyrimidin-7-one citrate salt, Revatio, UK-92,480
Empirical Formula (Hill Notation):
C22H30N6O4S ·C6H8O7
CAS Number:
Molecular Weight:
666.70
Products
Formula Weight
Gene Alias
Manufacturer
Format
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing