![Senecionine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/379/049/ebee9f7e-11d8-4fe2-92d6-c4f3b9db4b7e/640/ebee9f7e-11d8-4fe2-92d6-c4f3b9db4b7e.png)
Senecionine
![Senecionine phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/379/049/ebee9f7e-11d8-4fe2-92d6-c4f3b9db4b7e/640/ebee9f7e-11d8-4fe2-92d6-c4f3b9db4b7e.png)
Synonym(s):
Aureine
Empirical Formula (Hill Notation):
C18H25NO5
CAS Number:
Molecular Weight:
335.39
Products
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing