![(S)-(+)-Glutamine for synthesis](/deepweb/assets/sigmaaldrich/product/structures/100/376/398b026a-a679-409c-b93c-16abd79b23d6/640/398b026a-a679-409c-b93c-16abd79b23d6.png)
(S)-(+)-Glutamine
![(S)-(+)-Glutamine for synthesis](/deepweb/assets/sigmaaldrich/product/structures/100/376/398b026a-a679-409c-b93c-16abd79b23d6/640/398b026a-a679-409c-b93c-16abd79b23d6.png)
Synonym(s):
L-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, L-Glutamic acid 5-amide, Levoglutamide
Linear Formula:
H2NCOCH2CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
146.14
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1