Skip to Content
Merck
(S)-(+)-3-Hydroxytetrahydrofuran 99%

(S)-(+)-3-Hydroxytetrahydrofuran

(S)-(+)-3-Hydroxytetrahydrofuran 99%
Synonym(s):
(S)-(+)-Tetrahydro-3-furanol
Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing