(S)-(+)-1-Aminoethylphosphonic acid
Synonym(s):
D-(+)-1-Aminoethylphosphonic acid, D-Ala(P)
Linear Formula:
CH3CH(NH2)P(O)(OH)2
CAS Number:
Molecular Weight:
125.06
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing