Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Screw cap tubes and caps with O-rings, conical bottom
Products
Product Category
tubes (14)
Manufacturer
Capacity (mL)
Color
Package Size
Sterility
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
AXYST200CS
Screw cap tubes and caps with O-rings, conical bottom,
with O-ring, size 2.0 mL, pkg of 8x500caps/cs, clear, sterile
Expand
View Pricing
AXYSCT050OS
Screw cap tubes and caps with O-rings, conical bottom,
capacity 0.5 mL, orange, sterile
Expand
View Pricing
AXYSCT150OS
Screw cap tubes and caps with O-rings, conical bottom,
capacity 1.5 mL, orange, sterile
Expand
View Pricing
AXYSCT050RS
Screw cap tubes and caps with O-rings, conical bottom,
capacity 0.5 mL, red, sterile
Expand
View Pricing
AXYSCT050BS
Screw cap tubes and caps with O-rings, conical bottom,
capacity 0.5 mL, blue, sterile
Expand
View Pricing
AXYSCT050YS
Screw cap tubes and caps with O-rings, conical bottom,
capacity 0.5 mL, yellow, sterile
Expand
View Pricing
AXYSCT200BR
Screw cap tubes and caps with O-rings, conical bottom,
size 2.0 mL, brown, pkg of 8x500caps/cs
Expand
View Pricing
AXYST150CS
Screw cap tubes and caps with O-rings, conical bottom,
with O-ring, size 1.5 mL, pkg of 8x500caps/cs, clear
Expand
View Pricing
AXYSCT150VS
Screw cap tubes and caps with O-rings, conical bottom,
capacity 1.5 mL, violet, sterile
Expand
View Pricing
AXYSCT150BS
Screw cap tubes and caps with O-rings, conical bottom,
capacity 1.5 mL, blue, sterile
Expand
View Pricing
AXYSCT150GS
Screw cap tubes and caps with O-rings, conical bottom,
capacity 1.5 mL, green, sterile
Expand
View Pricing
AXYSCT150YS
Screw cap tubes and caps with O-rings, conical bottom,
capacity 1.5 mL, yellow, sterile
Expand
View Pricing
AXYSCT050GS
Screw cap tubes and caps with O-rings, conical bottom,
capacity 0.5 mL, green, sterile
Expand
View Pricing
AXYSCT150RS
Screw cap tubes and caps with O-rings, conical bottom,
capacity 1.5 mL, red, sterile
Expand
View Pricing