Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Salt Active Nuclease (SAN)
Products
Product Category
enzymes (3)
Brand
Sigma-Aldrich (4)
Biological Source
Physical Form
Recombinant Host
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
SRE0015
Salt Active Nuclease (SAN),
recombinant, expressed in
Pichia pastoris
Expand
View Pricing
SRE0115
Salt Active Nuclease (SAN),
Heat-labile, recombinant, expressed in
Pichia pastoris
, Triton-free
Expand
View Pricing
SRE0116
Salt Active Nuclease (SAN),
recombinant, expressed in
Pichia pastoris
, Triton-free
Expand
View Pricing
SRE0117
Salt Active Nuclease (SAN),
High Purity, recombinant, expressed in
Pichia pastoris
, Triton-free
Expand
View Pricing