Skip to Content
Merck
Saccharin Sodium Pharmaceutical Secondary Standard; Certified Reference Material

Saccharin Sodium

Saccharin Sodium Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Saccharin sodium, Saccharin sodium salt
Empirical Formula (Hill Notation):
C7H4NNaO3S
CAS Number:
Molecular Weight:
205.17

Products

Brand
Formula Weight
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1