
(S)-1-Boc-3-methylpiperazine

Synonym(s):
tert-Butyl (S)-3-methyl-1-piperazinecarboxylate
Empirical Formula (Hill Notation):
C10H20N2O2
CAS Number:
Molecular Weight:
200.28
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1