Rocuronium bromide
Synonym(s):
Rocuronium bromide
Empirical Formula (Hill Notation):
C32H53BrN2O4
CAS Number:
Molecular Weight:
609.68
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Package Size
API Family
Reference Material Type
Special Grade