(R)-(−)-Leucinol
Synonym(s):
(R)-2-Amino-4-methyl-1-pentanol, D-Leucinol
Linear Formula:
(CH3)2CHCH2CH(NH2)CH2OH
CAS Number:
Molecular Weight:
117.19
Products
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing