Skip to Content
Merck
Risedronate Impurity D Pharmaceutical Secondary Standard; Certified Reference Material

Risedronate Impurity D

Risedronate Impurity D Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
3-Pyridylacetic acid hydrochloride, 3-Pyridineacetic acid hydrochloride
Empirical Formula (Hill Notation):
C7H7NO2 · HCl
CAS Number:
Molecular Weight:
173.60

Products

Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1