Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Relative Density Standard (20°C)
EC Number:
232-455-8
Products
Manufacturer
Quality Segment
Application
Special Grade
certified reference material (5)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
RDEN2001
Relative Density Standard (20°C),
UKAS ISO/IEC17025 and ISO 17034 certified, Relative Density 20/4 0.6597 at 20 °C
Expand
View Pricing
RDEN2002
Relative Density Standard (20°C),
UKAS ISO/IEC17025 and ISO 17034 certified, Relative Density 20/4 0.7145 at 20 °C
Expand
View Pricing
RDEN2003
Relative Density Standard (20°C),
UKAS ISO/IEC17025 and ISO 17034 certified, Relative Density 20/4 0.7769 at 20 °C
Expand
View Pricing
RDEN2004
Relative Density Standard (20°C),
UKAS ISO/IEC17025 and ISO 17034 certified, Relative Density 20/4 0.8400 at 20 °C
Expand
View Pricing
RDEN2005
Relative Density Standard (20°C),
UKAS ISO/IEC17025 and ISO 17034 certified, Relative Density 20/4 0.8452 at 20 °C
Expand
View Pricing