
Rebaudioside B

Synonym(s):
(4α)-13-[(O-β-D-Glucopyranosyl-(1→2)-O-[β-D-glucopyranosyl-(1→3)]-β-D-glucopyranosyl)oxy]kaur-16-en-18-oic acid, Stevioside a4
Empirical Formula (Hill Notation):
C38H60O18
CAS Number:
Molecular Weight:
804.87
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1