![(R)-(−)-2-Amino-3-methylbutane ≥97.0% (GC)](/deepweb/assets/sigmaaldrich/product/structures/313/336/4140b6f3-1948-496e-8538-721b2e539713/640/4140b6f3-1948-496e-8538-721b2e539713.png)
(R)-(−)-2-Amino-3-methylbutane
![(R)-(−)-2-Amino-3-methylbutane ≥97.0% (GC)](/deepweb/assets/sigmaaldrich/product/structures/313/336/4140b6f3-1948-496e-8538-721b2e539713/640/4140b6f3-1948-496e-8538-721b2e539713.png)
Synonym(s):
(R)-(−)-3-Methyl-2-butylamine
Empirical Formula (Hill Notation):
C5H13N
CAS Number:
Molecular Weight:
87.16
Products
Product Category
Brand
Formula Weight
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing