Quercetin dihydrate
Synonym(s):
2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-1-benzopyran-4-one dihydrate, 3,3′,4′,5,7-Pentahydroxyflavone dihydrate
Empirical Formula (Hill Notation):
C15H10O7 · 2H2O
CAS Number:
Molecular Weight:
338.27
Products
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3