![Pyruvaldehyde solution 40 wt. % in H2O](/deepweb/assets/sigmaaldrich/product/structures/421/387/5e6912b9-e76d-44e0-a0fa-55ff18c2813c/640/5e6912b9-e76d-44e0-a0fa-55ff18c2813c.png)
Pyruvaldehyde solution
![Pyruvaldehyde solution 40 wt. % in H2O](/deepweb/assets/sigmaaldrich/product/structures/421/387/5e6912b9-e76d-44e0-a0fa-55ff18c2813c/640/5e6912b9-e76d-44e0-a0fa-55ff18c2813c.png)
Synonym(s):
Methylglyoxal solution, Acetylformaldehyde, Pyruvaldehyde, Pyruvic aldehyde
Linear Formula:
CH3COCHO
CAS Number:
Molecular Weight:
72.06
Products
Product Category
Brand
Biological Source
Formula Weight
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing