Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
ground-glass stopper
Synonym(s):
Corning
®
7575
Products
Product Category
stoppers (4)
Manufacturer
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CLS7570N24
Pyrex
®
ground-glass stopper,
joint: ST/NS 24/40, Full length, closed at bottom
Expand
View Pricing
CLS757514
Pyrex
®
ground-glass stopper,
joint: ST/NS 14/20, Combination reagent bottle/ground joint, hollow
Expand
View Pricing
CLS757524
Pyrex
®
ground-glass stopper,
joint: ST/NS 24/30, Combination reagent bottle/ground joint, hollow
Expand
View Pricing
CLS757519
Pyrex
®
ground-glass stopper,
joint: ST/NS 19/22, Combination reagent bottle/ground joint, hollow
Expand
View Pricing