Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
culture tubes, rimless
Synonym(s):
Corning
®
9820
Products
Product Category
tubes (12)
Color
Manufacturer
Capacity (mL)
OD (mm)
Cap
Package Size
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
CLS982025
Pyrex
®
culture tubes, rimless,
capacity 55 mL
Expand
View Pricing
CLS982016X
Pyrex
®
culture tubes, rimless,
capacity 15 mL
Expand
View Pricing
CLS982010
Pyrex
®
culture tubes, rimless,
capacity 3 mL
Expand
View Pricing
CLS982012
Pyrex
®
culture tubes, rimless,
capacity 5 mL
Expand
View Pricing
CLS98206
Pyrex
®
culture tubes, rimless,
capacity 0.5 mL
Expand
View Pricing
CLS982018
Pyrex
®
culture tubes, rimless,
capacity 27 mL
Expand
View Pricing
CLS982022
Pyrex
®
culture tubes, rimless,
capacity 50 mL
Expand
View Pricing
CLS982025X
Pyrex
®
culture tubes, rimless,
capacity 70 mL
Expand
View Pricing
CLS982013
Pyrex
®
culture tubes, rimless,
capacity 9 mL
Expand
View Pricing
CLS982016
Pyrex
®
culture tubes, rimless,
capacity 11 mL
Expand
View Pricing
CLS982020
Pyrex
®
culture tubes, rimless,
capacity 34 mL
Expand
View Pricing
CLS982016XX
Pyrex
®
culture tubes, rimless,
capacity 20 mL
Expand
View Pricing