Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
crystallizing dish
Synonym(s):
Corning
®
3140
Products
Product Category
dishes (8)
Manufacturer
Capacity (mL)
Height (mm)
OD (mm)
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
CLS3140190
Pyrex
®
crystallizing dish,
capacity 2,500 mL, O.D. × H ~190 mm × 100 mm
Expand
View Pricing
CLS3140125
Pyrex
®
crystallizing dish,
capacity 740 mL, O.D. × H ~125 mm × 65 mm
Expand
View Pricing
CLS3140150
Pyrex
®
crystallizing dish,
capacity 1,200 mL, O.D. × H ~150 mm × 75 mm
Expand
View Pricing
CLS3140170
Pyrex
®
crystallizing dish,
capacity 1,770 mL, O.D. × H ~170 mm × 90 mm
Expand
View Pricing
CLS314070
Pyrex
®
crystallizing dish,
capacity 180 mL, O.D. × H ~70 mm × 50 mm
Expand
View Pricing
CLS314080
Pyrex
®
crystallizing dish,
capacity 180 mL, O.D. × H ~80 mm × 40 mm
Expand
View Pricing
CLS314090
Pyrex
®
crystallizing dish,
capacity 270 mL, O.D. × H ~90 mm × 50 mm
Expand
View Pricing
CLS3140100
Pyrex
®
crystallizing dish,
capacity 325 mL, O.D. × H ~100 mm × 50 mm
Expand
View Pricing