
Pyrazole

Synonym(s):
1,2-Diazole, Pyrazole
Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2