Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ProteoMass
™
Insulin MALDI-MS Standard
Synonym(s):
Insulin from bovine pancreas
Empirical Formula (Hill Notation):
C
254
H
377
N
65
O
75
S
6
CAS Number:
11070-73-8
Molecular Weight:
5733.49
EC Number:
234-291-2
Products
Formula Weight
Quality Segment
Package Size
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
I6279
ProteoMass
™
Insulin MALDI-MS Standard,
vial of 10 nmol, monoisotopic mol wt 5,729.6087 Da
Expand
View Pricing