
Promethazine Related Compound B

Synonym(s):
Isopromethazine hydrochloride, N,N,β-Trimethyl-10H-phenothiazine-10-ethanamine monohydrochloride, N,N-Dimethyl-2-(10H-phenothiazin-10-yl)propan-1-amine hydrochloride
Empirical Formula (Hill Notation):
C17H20N2S · HCl
CAS Number:
Molecular Weight:
320.88
Products
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1