Prednisone for peak identification
Synonym(s):
Prednisone, 1,4-Pregnadiene-17α,21-diol-3,11,20-trione, 1-Cortisone, 17α,21-Dihydroxy-1,4-pregnadiene-3,11,20-trione, Dehydrocortisone
Empirical Formula (Hill Notation):
C21H26O5
CAS Number:
Molecular Weight:
358.43
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing