Skip to Content
Merck
Prednisolone Tebutate pharmaceutical secondary standard, certified reference material

Prednisolone tebutate

Prednisolone Tebutate pharmaceutical secondary standard, certified reference material
Synonym(s):
(11beta)-11,17-Dihydroxy-21-[(3,3-dimethyl-1-oxobutyl)oxy]pregna-1,4-diene-3,20-dione, Prednisolone 21-(3,3-dimethylbutyrate), Prednisolone tert-butylacetate
Empirical Formula (Hill Notation):
C27H38O6
CAS Number:
Molecular Weight:
458.59

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade