![Prednicarbate Related Compound C pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/365/596/16c4084e-5d30-49e7-9f15-b527ed025535/640/16c4084e-5d30-49e7-9f15-b527ed025535.jpg)
Prednicarbate Related Compound C
![Prednicarbate Related Compound C pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/365/596/16c4084e-5d30-49e7-9f15-b527ed025535/640/16c4084e-5d30-49e7-9f15-b527ed025535.jpg)
Synonym(s):
(11β)-11,17-Dihydroxy-21-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, Prednisolone 21-propionate
Empirical Formula (Hill Notation):
C24H32O6
CAS Number:
Molecular Weight:
416.51
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2