Prazosin hydrochloride
Synonym(s):
1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-(2-furanylcarbonyl)piperazine hydrochloride, Furazosin hydrochloride, Prazosin hydrochloride
Empirical Formula (Hill Notation):
C19H21N5O4 · HCl
CAS Number:
Molecular Weight:
419.86
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing