![Praziquantel Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/382/311/9b69eea5-3e36-4a5b-b778-003c37c07780/640/9b69eea5-3e36-4a5b-b778-003c37c07780.jpg)
Praziquantel Related Compound A
![Praziquantel Related Compound A pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/382/311/9b69eea5-3e36-4a5b-b778-003c37c07780/640/9b69eea5-3e36-4a5b-b778-003c37c07780.jpg)
Synonym(s):
(11bRS)-2-Benzoyl-1,2,3,6,7,11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one
Empirical Formula (Hill Notation):
C19H18N2O2
CAS Number:
Molecular Weight:
306.36
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2