Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Polysulfide in light mineral oil
CAS Number:
68425-16-1
Products
Manufacturer
Physical Form
Quality Segment
Special Grade
certified reference material (18)
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
LMPS170
Polysulfide in light mineral oil,
2.0000% (20000), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS040
Polysulfide in light mineral oil,
0.0050% (50 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS020
Polysulfide in light mineral oil,
0.0010% (10 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS060
Polysulfide in light mineral oil,
0.0250% (250 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS070
Polysulfide in light mineral oil,
0.0300% (300 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS080
Polysulfide in light mineral oil,
0.0500% (500 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS050
Polysulfide in light mineral oil,
0.0100% (100 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS120
Polysulfide in light mineral oil,
0.2500% (2500 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS130
Polysulfide in light mineral oil,
0.3000% (3000 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS150
Polysulfide in light mineral oil,
1.0000% (10000 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS160
Polysulfide in light mineral oil,
1.5000% (15000 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS180
Polysulfide in light mineral oil,
3.0000% (30000 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS030
Polysulfide in light mineral oil,
0.0025% (25 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS090
Polysulfide in light mineral oil,
0.0700% (700 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS110
Polysulfide in light mineral oil,
0.1000% (1000 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS140
Polysulfide in light mineral oil,
0.5000% (5000 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS010
Polysulfide in light mineral oil,
0.0005% (5 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing
LMPS100
Polysulfide in light mineral oil,
0.0750% (750 ppm), certified reference material (UKAS ISO 17025 & ISO 17034 certified)
Expand
View Pricing