Skip to Content
Products
Cart
0
AR
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Polystyrene-
block
-poly(ethylene-
ran
-butylene)-
block
-polystyrene
Linear Formula:
[-CH
2
CH(C
6
H
5
)-]
W
(-CH
2
CH
2
-)
x
[-CH
2
CH(C
2
H
5
)-]
y
[-CH
2
CH(C
6
H
5
)-]
Z
CAS Number:
66070-58-4
Products
Brand
Sigma-Aldrich (2)
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
200557
Polystyrene-
block
-poly(ethylene-
ran
-butylene)-
block
-polystyrene,
powder, average M
w
~118,000 by GPC, contains >0.03% antioxidant as inhibitor
Expand
View Pricing
200565
Polystyrene-
block
-poly(ethylene-
ran
-butylene)-
block
-polystyrene,
mol wt 65-115 kDa by GPC, powder (Coarse)
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
Español - ES
English - EN
Learn More