Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(lactide-
co
-glycolide)-
b
-poly(ethylene glycol)-
b
-poly(lactide-
co
-glycolide)
Linear Formula:
H((C
3
H
4
O
2
)
x
(C
2
H
2
O
2
)
y
)
m
((C
2
H
4
O)
z
)
n
O((C
3
H
4
O
2
)
x
(C
2
H
2
O
2
)
y
)
m
H
Products
Brand
Sigma-Aldrich (2)
Color
Physical Form
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
908843
Poly(lactide-
co
-glycolide)-
b
-poly(ethylene glycol)-
b
-poly(lactide-
co
-glycolide),
average M
n
(1,600-1,500-1,600), lactide:glycolide 75:25
Expand
View Pricing
909076
Poly(lactide-
co
-glycolide)-
b
-poly(ethylene glycol)-
b
-poly(lactide-
co
-glycolide),
average M
n
(1,700-1,500-1,700), lactide:glycolide (95:5)
Expand
View Pricing