Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(ethylene glycol) dimethyl ether
Synonym(s):
Dimethyl polyethylene glycol
Linear Formula:
CH
3
O(CH
2
CH
2
O)
n
CH
3
CAS Number:
24991-55-7
Products
Boiling Point (°C)
Physical Form
Functionality
Shape
Polymer Composition
Polymer Type
Reaction Type
Reagent Type
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
445886
Poly(ethylene glycol) dimethyl ether,
average M
N
500, cross-linking reagent click chemistry, 100 ppm BHT as stabilizer
Expand
View Pricing
445894
Poly(ethylene glycol) dimethyl ether,
average M
N
1,000, cross-linking reagent click chemistry
Expand
View Pricing
445878
Poly(ethylene glycol) dimethyl ether,
average M
N
250, cross-linking reagent click chemistry
Expand
View Pricing
445908
Poly(ethylene glycol) dimethyl ether,
average M
N
2,000, cross-linking reagent click chemistry
Expand
View Pricing