![Poly(ethylene glycol) diglycidyl ether Mn 6,000](/deepweb/assets/sigmaaldrich/product/structures/524/134/4a426b28-9302-430c-a7ad-7403ad71e8f3/640/4a426b28-9302-430c-a7ad-7403ad71e8f3.png)
Poly(ethylene glycol) diglycidyl ether
![Poly(ethylene glycol) diglycidyl ether Mn 6,000](/deepweb/assets/sigmaaldrich/product/structures/524/134/4a426b28-9302-430c-a7ad-7403ad71e8f3/640/4a426b28-9302-430c-a7ad-7403ad71e8f3.png)
Synonym(s):
Diepoxy PEG, PEG diglycidyl ether, Polyoxyethylene bis(glycidyl ether)
Linear Formula:
C3H5O2-(C2H4O)n-C3H5O
CAS Number:
Products
Brand
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Showing 1-3 of 3
Compare
Product Number
Description
Pricing