Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(3-hydroxybutyric acid)
Linear Formula:
(C
4
H
6
O
2
)
n
C
4
H
6
O
2
CAS Number:
26063-00-3
Products
Brand
Sigma-Aldrich (2)
Color
Physical Form
Quality Segment
Polymer Composition
Polymer Type
Special Grade
Natural (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
915092
Poly(3-hydroxybutyric acid),
natural origin, average M
n
~500,000
Expand
View Pricing
916358
Poly(3-hydroxybutyric acid),
average M
n
10,000
Expand
View Pricing