![Poloxamer 188 European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/320/746/253dda1a-6439-4d58-839b-4404693cf5f3/640/253dda1a-6439-4d58-839b-4404693cf5f3.png)
Poloxamer 188
![Poloxamer 188 European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/320/746/253dda1a-6439-4d58-839b-4404693cf5f3/640/253dda1a-6439-4d58-839b-4404693cf5f3.png)
Synonym(s):
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol)
Linear Formula:
(C3H6O · C2H4O)x
CAS Number:
Products
Product Category
Brand
Manufacturer
pH Value
Physical Form
Product Line
Agency
Format
Chemical Composition
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4