Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PLTMax Human Platelet Lysate
Products
Product Category
stem cells (1)
Brand
Sigma-Aldrich (2)
Biological Source
Physical Form
Quality Segment
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
SCM141
PLTMax Human Platelet Lysate,
PLTMax Human Platelet Lysate is a growth factor rich supplement that is a superior alternative to fetal bovine serum (FBS) for human mesenchymal stem cell (MSC) culture. 100ml.
Expand
View Pricing
SCM142
PLTMax Human Platelet Lysate,
PLTMax Human Platelet Lysate is a growth factor rich supplement that is a superior alternative to fetal bovine serum (FBS) for human mesenchymal stem cell (MSC) culture. 500ml.
Expand
View Pricing