Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platinum-rhodium alloy, Pt60Rh40
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
GF55258937
Platinum-rhodium alloy, Pt60Rh40,
1m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF67200094
Platinum-rhodium alloy, Pt60Rh40,
1m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF92201756
Platinum-rhodium alloy, Pt60Rh40,
0.1m wire, 0.8mm diameter, as drawn
Expand
View Pricing
GF09755477
Platinum-rhodium alloy, Pt60Rh40,
2m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF73445373
Platinum-rhodium alloy, Pt60Rh40,
0.2m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF99982771
Platinum-rhodium alloy, Pt60Rh40,
0.5m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF26200482
Platinum-rhodium alloy, Pt60Rh40,
0.2m wire, 0.8mm diameter, as drawn
Expand
View Pricing
GF19599744
Platinum-rhodium alloy, Pt60Rh40,
0.05m wire, 0.8mm diameter, as drawn
Expand
View Pricing
GF83274150
Platinum-rhodium alloy, Pt60Rh40,
0.1m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF03540697
Platinum-rhodium alloy, Pt60Rh40,
0.5m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF28103712
Platinum-rhodium alloy, Pt60Rh40,
5m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF65722541
Platinum-rhodium alloy, Pt60Rh40,
0.05m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF92541641
Platinum-rhodium alloy, Pt60Rh40,
2m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF47036910
Platinum-rhodium alloy, Pt60Rh40,
0.1m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF00527880
Platinum-rhodium alloy, Pt60Rh40,
0.2m wire, 0.125mm diameter, as drawn
Expand
View Pricing