Skip to Content
Merck
Pindolol pharmaceutical secondary standard, certified reference material

Pindolol

Pindolol pharmaceutical secondary standard, certified reference material
Synonym(s):
1-(1H-Indol-4-yloxy)-3-(isopropylamino)-2-propanol, 1-(1H-Indol-4-yloxy)-3-[(1-methylethyl)amino]-2-propanol, Pindolol
Empirical Formula (Hill Notation):
C14H20N2O2
CAS Number:
Molecular Weight:
248.32

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade