![Piceatannol powder](/deepweb/assets/sigmaaldrich/product/structures/419/609/7a5a1566-cb1b-44dd-92d2-c4f8c0fcf1c9/640/7a5a1566-cb1b-44dd-92d2-c4f8c0fcf1c9.png)
Piceatannol
![Piceatannol powder](/deepweb/assets/sigmaaldrich/product/structures/419/609/7a5a1566-cb1b-44dd-92d2-c4f8c0fcf1c9/640/7a5a1566-cb1b-44dd-92d2-c4f8c0fcf1c9.png)
Synonym(s):
(E)-4-[2-(3,5Dihydroxyphenyl)ethenyl]1,2-benzenediol, 3,3′,4,5′-Stilbenetetrol, 3,3′,4,5′-Tetrahydroxy-trans-stilbene, Astringenin
Empirical Formula (Hill Notation):
C14H12O4
CAS Number:
Molecular Weight:
244.24
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing