![Phthalimide-15N potassium salt 98 atom % 15N](/deepweb/assets/sigmaaldrich/product/structures/204/945/423aa12e-25e4-4c2e-a0f7-0cf6e1db438f/640/423aa12e-25e4-4c2e-a0f7-0cf6e1db438f.png)
Phthalimide-15N potassium salt
![Phthalimide-15N potassium salt 98 atom % 15N](/deepweb/assets/sigmaaldrich/product/structures/204/945/423aa12e-25e4-4c2e-a0f7-0cf6e1db438f/640/423aa12e-25e4-4c2e-a0f7-0cf6e1db438f.png)
Synonym(s):
1,3-Dihydro-1,3-dioxoisoindole-15N, Potassium phthalimide-15N
Empirical Formula (Hill Notation):
C8H4K15NO2
CAS Number:
Molecular Weight:
186.21
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing