Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PHTHALIC ACID MONO-(1-METHYL-2-PHENYL-PROPYL) ESTER
Linear Formula:
C
18
H
18
O
4
CAS Number:
856806-12-7
Molecular Weight:
298.342
Products
Formula Weight
Special Grade
Aldrich
CPR
(9)
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
S13111
PHTHALIC ACID MONO-(1-METHYL-2-PHENYL-PROPYL) ESTER,
Aldrich
CPR
Expand
View Pricing
S334332
PHTHALIC ACID MONO-(1-METHYL-2-PHENYL-PROPYL) ESTER,
Aldrich
CPR
Expand
View Pricing
S334502
PHTHALIC ACID MONO-(1-METHYL-2-PHENYL-PROPYL) ESTER,
Aldrich
CPR
Expand
View Pricing
S484970
PHTHALIC ACID MONO-(1-METHYL-2-PHENYL-PROPYL) ESTER,
Aldrich
CPR
Expand
View Pricing
S484857
PHTHALIC ACID MONO-(1-METHYL-2-PHENYL-PROPYL) ESTER,
Aldrich
CPR
Expand
View Pricing
S313572
PHTHALIC ACID MONO-(1-METHYL-2-PHENYL-PROPYL) ESTER,
Aldrich
CPR
Expand
View Pricing
S339806
PHTHALIC ACID MONO-(1-METHYL-2-PHENYL-PROPYL) ESTER,
Aldrich
CPR
Expand
View Pricing
S313564
PHTHALIC ACID MONO-(1-METHYL-2-PHENYL-PROPYL) ESTER,
Aldrich
CPR
Expand
View Pricing
S490482
PHTHALIC ACID MONO-(1-METHYL-2-PHENYL-PROPYL) ESTER,
Aldrich
CPR
Expand
View Pricing