Phillyrin
Synonym(s):
Forsythin, Phillyroside, Phyllyrin
Empirical Formula (Hill Notation):
C27H34O11
CAS Number:
Molecular Weight:
534.55
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing