Skip to Content
Merck
vibrant-m

Pheniramine maleate

vibrant-m
Synonym(s):
Pheniramine maleate salt
Empirical Formula (Hill Notation):
C16H20N2 · C4H4O4
CAS Number:
Molecular Weight:
356.42

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade