Skip to Content
Merck
Phenformin Hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material

Phenformin hydrochloride

Phenformin Hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
N-(2-Phenylethyl)imidodicarbonimidic diamide monohydrochloride, Phenethylbiguanide, 1-(Diaminomethylidene)-2-(2-phenylethyl)guanidine hydrochloride, 1-Carbamimidamido-N-(2-phenylethyl)methanimidamide hydrochloride, 1-Phenethyl-biguanide hydrochloride, N-(2-Phenylethyl)imidodicarbonimidic diamide hydrochloride, Phenformin hydrochloride
Empirical Formula (Hill Notation):
C10H15N5 · HCl
CAS Number:
Molecular Weight:
241.72
EC Number:

Products

Brand
Color
Formula Weight
Physical Form
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique