Skip to Content
Merck
p-Fluorobutyryl fentanyl (PFBF) solution 100 μg/mL in methanol (), certified reference material, ampule of 0.5 mL, Cerilliant®

p-Fluorobutyryl fentanyl (PFBF) solution

p-Fluorobutyryl fentanyl (PFBF) solution 100 μg/mL in methanol (), certified reference material, ampule of 0.5 mL, Cerilliant®
Empirical Formula (Hill Notation):
C23H29FN2O
CAS Number:
Molecular Weight:
368.49

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique